missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAB3 Recombinant Rabbit Monoclonal Antibody (24GB2680)

Rabbit Recombinant Monoclonal Antibody
£126.00 - £434.00
Specifications
| Antigen | TAB3 |
|---|---|
| Clone | 24GB2680 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30276649
![]() |
Invitrogen™
MA557277 |
100 μL |
£434.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30276119
|
Invitrogen™
MA557278 |
20 μL |
£126.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TAB3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
TAB3 functions in the NF-kappaB signal transduction pathway. It and the similar and functionally redundant protein TAB2, form a ternary complex with the protein kinase TAK1 and either TRAF2 or TRAF6 in response to stimulation with the pro-inflammatory cytokines TNF or IL-1. Subsequent TAK1 kinase activity triggers a signaling cascade leading to activation of the NF-kappaB transcription factor. Recent experiments have shown that TAB2 and the related protein TAB3 constitutitvely interact with the autophagy mediator Beclin-1; upon induction of autophagy, these proteins dissociate from Beclin-1 and bind TAK1. Overexpression of TAB2 and TAB3 inhibit autophagy, while their depletion triggers it, suggesting that TAB2 and TAB3 act as a control point for autophagy.Specifications
| TAB3 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| Tab3 | |
| 4921526G09Rik; Kiaa4135; MAP3K7IP3; mitogen activated protein kinase kinase kinase 7 interacting protein 3; mitogen-activated protein kinase kinase kinase 7 interacting protein 3; mitogen-activated protein kinase kinase kinase 7-interacting protein 3; mKIAA4135; NAP1; NF-kappa-B-activating protein 1; NFkB activating protein 1; Tab3; TAB-3; TAK1-binding protein 3; TGF-beta activated kinase 1 (MAP3K7) binding protein 3; TGF-beta activated kinase 1 and MAP3K7 binding protein 3; TGF-beta activated kinase 1/MAP3K7 binding protein 3; TGF-beta-activated kinase 1 and MAP3K7-binding protein 3; TGF-beta-activated kinase 1-binding protein 3 | |
| Tab3 | |
| Primary | |
| Affinity chromatography |
| 24GB2680 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q571K4, Q8N5C8 | |
| 257397, 66724 | |
| A synthesized peptide derived from human TAB3 (aa 50-94). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title