missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAB2 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81735AF350
This item is not returnable.
View return policy
Description
TAB2 Polyclonal antibody specifically detects TAB2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| TAB2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 350 | |
| FLJ21885, KIAA0733CHTD2, MAP3K7IP2mitogen-activated protein kinase kinase kinase 7 interacting protein 2, Mitogen-activated protein kinase kinase kinase 7-interacting protein 2, TAB-2, TAK1-binding protein 2, TGF-beta activated kinase 1/MAP3K7 binding protein 2, TGF-beta-activated kinase 1 and MAP3K7-binding protein 2, TGF-beta-activated kinase 1-binding protein 2 | |
| TAB2 antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human TAB2. The immunogen is located within the last 50 amino acids of TAB2. Amino Acid Squence: KTPKTQDTEDDEGA | |
| 0.1 mL | |
| Cytokine Research, Signal Transduction | |
| 23118 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction