missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAB1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-21405
This item is not returnable.
View return policy
Description
TAB1 Polyclonal specifically detects TAB1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| TAB1 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| MAP3K7IP13'-Tab1, MGC57664, mitogen-activated protein kinase kinase kinase 7 interacting protein 1, Mitogen-activated protein kinase kinase kinase 7-interacting protein 1, TAK1-binding protein 1, TGF-beta activated kinase 1/MAP3K7 binding protein 1, TGF-beta-activated kinase 1 and MAP3K7-binding protein 1, TGF-beta-activated kinase 1-binding protein 1, transforming growth factor beta-activated kinase-binding protein 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate | |
| NP_006107.1 | |
| TAB1 | |
| The immunogen recognized by this antibody maps to a region between residue 435 and 485 of human TAK1-binding protein 1 using the numbering given in entry NP_006107.1 (GeneID 10454). | |
| 0.1 mL | |
| Immunology, Innate Immunity | |
| 10454 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction