missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAB1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£153.00
Specifications
| Antigen | TAB1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5-2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1-2 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
TAB1 Polyclonal specifically detects TAB1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| TAB1 | |
| Western Blot 0.5-2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1-2 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Immunology, Innate Immunity | |
| PBS with 0.02% Sodium Azide | |
| MAP3K7IP13'-Tab1, MGC57664, mitogen-activated protein kinase kinase kinase 7 interacting protein 1, Mitogen-activated protein kinase kinase kinase 7-interacting protein 1, TAK1-binding protein 1, TGF-beta activated kinase 1/MAP3K7 binding protein 1, TGF-beta-activated kinase 1 and MAP3K7-binding protein 1, TGF-beta-activated kinase 1-binding protein 1, transforming growth factor beta-activated kinase-binding protein 1 | |
| TAB1 | |
| IgG | |
| Affinity Purified |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| NP_006107 | |
| 10454 | |
| Antibody was raised against a synthetic peptide corresponding to 13 amino acids in the center of human TAB1. The immunogen is located within amino acids 220 - 270 of TAB1. Amino Acid Squence: ESTRRIGDYKVKY | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title