missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAAR9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA132844
This item is not returnable.
View return policy
Description
PA1-32844 detects Trace Amine receptor 3 from human samples.
TAAR9 is a member of a large family of rhodopsin G protein-coupled receptors (GPCRs, or GPRs). GPCRs contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins.
Specifications
| TAAR9 | |
| Polyclonal | |
| Unconjugated | |
| TAAR9 | |
| Gm231; mTaar9; SNORF1 receptor; TA3; Taar9; TAR3; TaR-3; TAR9; taR-9; trace amine associated receptor 9 (gene/pseudogene); trace amine receptor 3; trace amine receptor 9; trace amine-associated receptor 9; trace-amine-associated receptor 9; Trar3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 134860 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q96RI9 | |
| TAAR9 | |
| Synthetic 12 amino acid peptide from 2nd extracellular domain of human TA3 | |
| 25 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto