missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAAR8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534243
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla (100%) Elephant (94%) Panda (81%).
Trace amines are biogenic amines present in very low levels in mammalian tissues. Although some trace amines have clearly defined roles as neurotransmitters in invertebrates, the extent to which they function as true neurotransmitters in vertebrates has remained speculative. Trace amines are likely to be involved in a variety of physiological functions that have yet to be fully understood. Expressed in kidney and amygdala. Not expressed in other tissues or brain regions tested.
Specifications
| TAAR8 | |
| Polyclonal | |
| Unconjugated | |
| TAAR8 | |
| G protein-coupled receptor 102; Gm1149; Gm230; GPR102; G-protein coupled receptor 102; mTaar8a; Q923; Ta10; Ta11; TA5; Ta7; TAAR8; Taar8a; Taar8b; Taar8c; Tar11; TaR-11; TAR5; TaR-5; TaR-8; TaR-8a; trace amine associated receptor 8; trace amine receptor 11; Trace amine receptor 5; trace amine receptor 8; trace amine receptor 8a; trace amine-associated receptor 8; Trace amine-associated receptor 8a; trace-amine-associated receptor 8a; Trar11; Trar5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 83551 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q969N4 | |
| TAAR8 | |
| Synthetic 16 amino acid peptide from 3rd cytoplasmic domain of human TAAR8. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction