missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAAR8 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NLS2307
This item is not returnable.
View return policy
Description
TAAR8 Polyclonal antibody specifically detects TAAR8 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| TAAR8 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 4 μg/mL | |
| GPR102G protein-coupled receptor 102, TA5TaR-5, TAR5G-protein coupled receptor 102, trace amine associated receptor 8, Trace amine receptor 5TaR-8, Trace amine receptor 8, trace amine-associated receptor 8, TRAR5 | |
| Synthetic 16 amino acid peptide from 3rd cytoplasmic domain of human TAAR8. | |
| 0.05 mL | |
| GPCR | |
| 83551 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction