missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAAR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595704
This item is not returnable.
View return policy
Description
Immunogen sequence: TSTDIMLSSA SIFHLSFISI DRYYAVCDPL RYKAKMNILV ICVMIFISWS VPAVFAFGMI FLELNFKGAE EIYYKHVHCR GGCSVFFSKI SGVLTFMTSF Y; Positive Samples: A-549, Rat stomach; Cellular Location: Cell membrane, Multi-pass membrane protein.
TA1 has been reported in human brain, dorsal root ganglion, olfactory bulb, kidney, liver, lung, pancreas, prostate, skeletal muscle, small intestine, spleen, spinal cord, and stomach. An EST for TA1 has been identified from a human stomach cancer library. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more than a thousand members currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This turn-off activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates.
Specifications
| TAAR1 | |
| Polyclonal | |
| Unconjugated | |
| Taar1 | |
| MGC126874; MGC138399; RP11-295F4.9; RTAR1; Snorf33; Ta1; Taar1; TAR1; taR-1; trace amine associated receptor 1; trace amine receptor 1; trace amine-associated receptor 1; trace-amine-associated receptor 1; TRAR1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 111174, 113914, 134864 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.07 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q923Y8, Q923Y9, Q96RJ0 | |
| Taar1 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human TAAR1 (NP_6122001). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction