missing translation for 'onlineSavingsMsg'
Learn More

(-)-Ketoconazole, MedChemExpress

Product Code. 16424237
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16424237

Brand: Medchem Express HYB0105B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (-)-Ketoconazole ((-)-R 41400) is one of the enantiomer of Ketoconazole. Ketoconazole is a racemic mixture of two enantiomers, levoketoconazole ((2S,4R)-(−)-ketoconazole) and dextroketoconazole ((2R,4S)-(+)-ketoconazole).

(-)-Ketoconazole ((-)-R 41400) is one of the enantiomer of Ketoconazole. Ketoconazole is a racemic mixture of two enantiomers, levoketoconazole ((2S,4R)-(−)-ketoconazole) and dextroketoconazole ((2R,4S)-(+)-ketoconazole).

Specifications

Chemical Name or Material (-)-Ketoconazole
CAS 142128-57-2
Color White
Purity Grade Notes Research
Molecular Formula C26H28Cl2N4O4
Quantity 50 mg
Synonym (-)-Ketoconazol (-)-R 41400
Solubility Information DMSO : 33.33 mg/mL (62.72 mM; Need ultrasonic)
SMILES CC(N1CCN(C2=CC=C(OC[C@H]3O[C@](CN4C=CN=C4)(C5=CC=C(Cl)C=C5Cl)OC3)C=C2)CC1)=O
Molecular Weight (g/mol) 531.43
Formula Weight 531.43
Percent Purity 99.71%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.