missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ T7 RNA Polymerase, in 20mM Potassium Phosphate and 10mM DTT Kit

Product Code. 15467119
Click to view available options
Quantity:
1 KU
Unit Size:
1kg
This item is not returnable. View return policy

Product Code. 15467119

Brand: Alfa Aesar™ J65896.4I

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For synthesis of RNA transcripts, biologically active mRNA and antisense RNA

TRUSTED_SUSTAINABILITY

Specifications

CAS 9014-24-8
Color Colorless
Content And Storage -30°C to -10°C
For Use With (Application) For synthesis of RNA transcripts, biologically active mRNA and antisense RNA
Product Type RNA Polymerase Kit
Quantity 1 KU
Synonym EC 2.7.7.6
Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.