missing translation for 'onlineSavingsMsg'
Learn More

T4-ATA (S-isomer), MedChemExpress

Product Code. p-200101326
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16461209

Brand: Medchem Express HY114272A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress T4-ATA S-isomer is the S-isomer of T4-ATA, which is the active form of the thyroid hormone.

T4-ATA S-isomer is the S-isomer of T4-ATA, which is the active form of the thyroid hormone.

Specifications

Chemical Name or Material T4-ATA (S-isomer)
CAS 2448470-97-9
Color White
Purity Grade Notes Research
Molecular Formula C19H15I4NO6S
Quantity 1 mg
SMILES CC(SCC(N[C@H](C(O)=O)CC(C=C1I)=CC(I)=C1OC2=CC(I)=C(O)C(I)=C2)=O)=O
Molecular Weight (g/mol) 893.01
Formula Weight 893.01
Percent Purity 98.1%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.