missing translation for 'onlineSavingsMsg'
Learn More

T4-ATA (S-isomer), MedChemExpress

Product Code. 16461209
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16461209 1 mg 1mg
16444808 10 mg 10mg
16417557 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16461209 Supplier Medchem Express Supplier No. HY114272A
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress T4-ATA S-isomer is the S-isomer of T4-ATA, which is the active form of the thyroid hormone.

T4-ATA S-isomer is the S-isomer of T4-ATA, which is the active form of the thyroid hormone.

Specifications

Chemical Name or Material T4-ATA (S-isomer)
CAS 2448470-97-9
Color White
Purity Grade Notes Research
Molecular Formula C19H15I4NO6S
Quantity 1 mg
SMILES CC(SCC(N[C@H](C(O)=O)CC(C=C1I)=CC(I)=C1OC2=CC(I)=C(O)C(I)=C2)=O)=O
Molecular Weight (g/mol) 893.01
Formula Weight 893.01
Percent Purity 98.1%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.