missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ T4

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
T4 is a modulator of alternative polyadenylation (APA) (IC50 values are 2.1 μM and 8.5 μM in U2OS and 293T cells, respectively). T4 promotes distal-to-proximal APA usage in multiple transcripts, using distinctive A-rich motifs through autoregulated PABPN1 signaling. T4 exhibits no inhibitory activity against a panel of 414 kinases. T4 prevents retinal degeneration in the RhoP23H mouse model of retinitis pigmentosa. T4 inhibits rod photoreceptor gene expression via Nr2e3 in a luciferase-based assay (IC50 = 0.07 μM).
Specifications
Specifications
| Chemical Name or Material | 6,7-Dimethoxy-2-[((4-phenyl-3,6-dihydropyridin-1(2 H)-yl)methyl)]quinazolin-4(3 H)-one |
| CAS | 785708-33-0 |
| Quantity | 50 mg |
| Target | DNA, RNA and Protein Synthesis Compounds |
| Molecular Formula | C22H23N3O3 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction