missing translation for 'onlineSavingsMsg'
Learn More
Learn More
T3-ATA (S-isomer), MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Description
T3-ATA S-isomer is the S-isomer of T3-ATA, which is the active form of the thyroid hormone.
Specifications
Specifications
| Chemical Name or Material | T3-ATA (S-isomer) |
| CAS | 2438721-48-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C19H16I3NO6S |
| Quantity | 5 mg |
| SMILES | CC(SCC(N[C@H](C(O)=O)CC(C=C1I)=CC(I)=C1OC2=CC=C(O)C(I)=C2)=O)=O |
| Molecular Weight (g/mol) | 767.11 |
| Formula Weight | 767.11 |
| Percent Purity | 99.5% |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction