missing translation for 'onlineSavingsMsg'
Learn More

T025, MedChemExpress

Product Code. 16477327
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477327 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16477327 Supplier Medchem Express Supplier No. HY112296

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress T025 is an orally active and highly potent inhibitor of Cdc2-like kinase (CLKs), with Kd values of 4.8, 0.096, 6.5, 0.61, 0.074, 1.5 and 32 nM for CLK1, CLK2, CLK3, CLK4, DYRK1A, DYRK1B and DYRK2, respectively.

T025 induces caspase-3/7-mediated cell apoptosis. T025 reduces CLK-dependent phosphorylation. T025 exerts anti-proliferative activities in both hematological and solid cancer cell lines (IC50 values: 30-300 nM). T025 has an anti-tumor efficiency, mainly for MYC-driven disease research.

Specifications

Chemical Name or Material T025
Physical Form Solid
CAS 2407433-00-3
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C21H18N8
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 8.25 mg/mL (21.57 mM; Need ultrasonic)
SMILES CNC1=NC(NCC2=NC=CC=N2)=C(C(C3=CC=C(N=CC=C4)C4=C3)=CN5)C5=N1
Molecular Weight (g/mol) 382.42
Formula Weight 382.42
Percent Purity 98.01%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.