missing translation for 'onlineSavingsMsg'
Learn More

T0070907, MedChemExpress

Product Code. 16465548
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16465548 10 mg 10mg
16456106 100 mg 100mg
16451717 50 mg 50mg
16438257 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16465548 Supplier Medchem Express Supplier No. HY13202
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress T0070907 is a potent PPARγ antagonist with a Ki of 1 nM.

T0070907 is a potent PPARγ antagonist with a Ki of 1 nM.

Specifications

Chemical Name or Material T0070907
CAS 313516-66-4
Color Light Nude
Purity Grade Notes Research
Molecular Formula C12H8ClN3O3
Quantity 10 mg
Solubility Information DMSO : 62.5 mg/mL (225.10 mM; ultrasonic and warming and heat to 60°C) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES ClC1=C(C=C(C=C1)[N+]([O-])=O)C(NC2=CC=NC=C2)=O
Molecular Weight (g/mol) 277.66
Formula Weight 277.66
Percent Purity 99.54%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.