missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ T-Shape Tubing Connector

Product Code. 15261509 Shop All Brand Products
Click to view available options
For Use With:
14-15 mm ID Tubing
Packaging Quantity:
10/Pk.
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 15261509

Brand: BRAND™ 152825

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Manufactured from translucent polypropylene, Brand™ T-Shape Tubing Connectors can be repeatedly autoclaved at 121°C.

  • Good chemical resistance and mechanical properties
  • Straight-through tube and side arm
  • Provided in packs of 20 (152815) and 10 (152825)

TRUSTED_SUSTAINABILITY

Specifications

Description Length of straight-through tube: 89mm; length of side arm including diameter of straight-through tube: 60mm
Autoclavable Autoclavable
Material Polypropylene
For Use With 14-15 mm ID Tubing
Packaging Quantity 10/Pk.
Color Translucent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.