missing translation for 'onlineSavingsMsg'
Learn More

t-Plasminogen Activator/tPA Antibody [HRP], Novus Biologicals™

Product Code. 30738151 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30738151 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30738151 Supplier Novus Biologicals Supplier No. AF7449H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

t-Plasminogen Activator/tPA Polyclonal antibody specifically detects t-Plasminogen Activator/tPA in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen t-Plasminogen Activator/tPA
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias Alteplase, DKFZp686I03148, EC 3.4.21, EC 3.4.21.68, plasminogen activator, tissue, plasminogen activator, tissue type, reteplase, tissue plasminogen activator (t-PA), tissue-type plasminogen activator, TPA, T-PA, t-plasminogen activator
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human t-Plasminogen Activator/tPA, Ser36-Pro562, Accession # P00750
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cell Biology, Endocrinology
Primary or Secondary Primary
Gene ID (Entrez) 5327
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.