missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ T Phosphoramidite, standard grade, serum vial bottle

Product Code. 18044070 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 18044070

Brand: Thermo Scientific™ 27173604

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

T CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxythymidine (dT) for use in oligonucleotide synthesis.

T CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxythymidine (dT) for use in oligonucleotide synthesis. DNA phosphoramidites are the building blocks for synthesis of DNA or DNA containing oligonucleotides. Each base has unique protecting groups (except T and U) designed specifically to protect the DNA during the chemical synthesis process, and produce the highest amount of full length product. Alternative protecting groups on dC (acetyl) and dG (DMF) allow for a faster deprotection of the oligonucleotide. The 'Standard Grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in a 20 mL Amber bottle with a Serum Vial neck style, which is compatible with most ABI Systems|AKTA Systems of oligonucleotide synthesizers.

Features:
Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-2'-deoxythymidine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
Molecular formula: C40H49N4O8P
Base: T
Formula weight: 744.82 g/mol
CAS number: 98796-51-1
HPLC Purity: ≥98.0%
31P NMR Purity: ≥98.0%

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) AKTA Synthesizers, Applied Biosystems Synthesizers
Product Type T CEP
Content And Storage Store in freezer (at or below -20°C).
Nucleoside Type Deoxythymidine (dT)
Shipping Condition Room Temperature
Purity 98%
Quantity 1 g
Reagent Type Phosphoramidite
Grade Standard
Nucleic Acid Type DNA
Form Powder
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.