Learn More
Thermo Scientific™ T Phosphoramidite, standard grade, serum vial bottle
Shop All Thermo Scientific ProductsDescription
T CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxythymidine (dT) for use in oligonucleotide synthesis. DNA phosphoramidites are the building blocks for synthesis of DNA or DNA containing oligonucleotides. Each base has unique protecting groups (except T and U) designed specifically to protect the DNA during the chemical synthesis process, and produce the highest amount of full length product. Alternative protecting groups on dC (acetyl) and dG (DMF) allow for a faster deprotection of the oligonucleotide. The 'Standard Grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in a 20 mL Amber bottle with a Serum Vial neck style, which is compatible with most ABI Systems|AKTA Systems of oligonucleotide synthesizers.
Features:
• Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-2'-deoxythymidine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula: C40H49N4O8P
• Base: T
• Formula weight: 744.82 g/mol
• CAS number: 98796-51-1
• HPLC Purity: ≥98.0%
• 31P NMR Purity: ≥98.0%
Specifications
Specifications
| For Use With (Equipment) | AKTA Synthesizers, Applied Biosystems Synthesizers |
| Product Type | T CEP |
| Content And Storage | Store in freezer (at or below -20°C). |
| Nucleoside Type | Deoxythymidine (dT) |
| Shipping Condition | Room Temperature |
| Purity | 98% |
| Quantity | 5 g |
| Reagent Type | Phosphoramidite |
| Grade | Standard |
| Nucleic Acid Type | DNA |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.