missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ T-Lymphocytes Monoclonal Antibody (KEN-5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA528508
This item is not returnable.
View return policy
Description
Mouse anti Rabbit T lymphocytes, clone KEN-5, recognizes rabbit (lapine) T-cells.
T Lymphocytes are a subtype of white blood cell that is central to cell-mediated immunity. T Lymphocytes mature in the thymus from thymocytes. The majority of human T cells rearrange their alpha and beta chains on the cell receptor and are termed alpha beta T cells (alpha/beta T cells) and are part of the adaptive immune system.
Specifications
| T-Lymphocytes | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| Rabbit Thymocytes. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
| KEN-5 | |
| Unconjugated | |
| Mouse | |
| Protein A | |
| RUO | |
| Rabbit | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction