Learn More
T Cell Exhaustion Marker (Flow) Antibody Pack, Novus Biologicals™
Antibody Pack
Brand:  Novus Biologicals NBP3-12864
Product Code. 18302614
  • £907.00 / Pack of 9
Includes: F0102B: Goat anti-Mouse IgG Secondary Antibody [Phycoerythrin], F0110: Goatanti-Rabbit IgG Secondary Antibody [Phycoerythrin], F0105B: Goat anti-Rat IgGSecondary Antibody [Phycoerythrin], MAB10863-SP, MAB3253-SP, MAB7898-SP, MAB2365R-SP, MAB71263-SP, MAB23191-SP
Description
Description
The Human T Cell Exhaustion Marker Sample Pack (Flow) contains antibodies to detect expression of inhibitory receptors associated with T cell exhaustion including PD-1, CTLA-4, TIGIT, TIM-3, LAG-3 and VISTA. Immune cells expressing these inhibitory receptors are found during chronic viral infection and within the tumor microenvironment of many solid tumors. Also included are three PE-Conjugated secondary antibodies for fluorescent detection by flow cytometry.All antibodies have been validated for use in flow cytometry applications.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
T Cell Exhaustion Marker (Flow)
Unconjugated
exhaustion markers, immune cell exhaustion, immune checkpoint markers, inhibitory receptors, T cell exhaustion
9 Vials
Storage is content dependent.
Flow Cytometry
See individual datasheets
See individual datasheets