missing translation for 'onlineSavingsMsg'
Learn More

t-Butyl4-[4-(4,4,5,5-teramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazinecarboxylate, 90%, Thermo Scientific™

£33.40 - £327.00

Chemical Identifiers

CAS 470478-90-1
Molecular Formula C21H33BN2O4
Molecular Weight (g/mol) 388.32
MDL Number MFCD06411544
InChI Key ZMAVVXGWEHZLDW-UHFFFAOYSA-N
Synonym tert-butyl 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazine-1-carboxylate, 4-4-boc-piperazino phenylboronic acid pinacol ester, 4-4-n-boc piperazin-1-yl phenylboronic acid pinacol ester, 4-4-tert-butoxycarbonylpiperazinyl phenylboronic acid pinacol ester, 4-4-tert-butoxycarbonylpiperazinyl phenylboronic acid, pinacol ester, tert-butyl 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl tetrahydro-1 2h-pyrazinecarboxylate, 4-4-boc-piperazinyl phenylboronic acid pinacol ester, 4-n-tert-butoxycarbonylpiperazine benzeneboronic acid pinacol ester, tert-butyl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazine-1-carboxylate
PubChem CID 2795514
IUPAC Name tert-butyl 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10740374
View Documents Promotion Details
Thermo Scientific Maybridge
CC30039CB
250 mg Amber Glass Bottle
£33.40
250mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10361013
View Documents Promotion Details
Thermo Scientific Maybridge
CC30039DA
1 g Amber Glass Bottle
£70.20
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11376825
View Documents Promotion Details
Thermo Scientific Maybridge
CC30039DE
5 g Amber Glass Bottle
£327.00
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

470478-90-1
388.32
ZMAVVXGWEHZLDW-UHFFFAOYSA-N
2795514
CC(C)(C)OC(=O)N1CCN(CC1)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
C21H33BN2O4
MFCD06411544
tert-butyl 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazine-1-carboxylate, 4-4-boc-piperazino phenylboronic acid pinacol ester, 4-4-n-boc piperazin-1-yl phenylboronic acid pinacol ester, 4-4-tert-butoxycarbonylpiperazinyl phenylboronic acid pinacol ester, 4-4-tert-butoxycarbonylpiperazinyl phenylboronic acid, pinacol ester, tert-butyl 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl tetrahydro-1 2h-pyrazinecarboxylate, 4-4-boc-piperazinyl phenylboronic acid pinacol ester, 4-n-tert-butoxycarbonylpiperazine benzeneboronic acid pinacol ester, tert-butyl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazine-1-carboxylate
tert-butyl 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazine-1-carboxylate

Specifications

470478-90-1
C21H33BN2O4
tert-butyl 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazine-1-carboxylate, 4-4-boc-piperazino phenylboronic acid pinacol ester, 4-4-n-boc piperazin-1-yl phenylboronic acid pinacol ester, 4-4-tert-butoxycarbonylpiperazinyl phenylboronic acid pinacol ester, 4-4-tert-butoxycarbonylpiperazinyl phenylboronic acid, pinacol ester, tert-butyl 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl tetrahydro-1 2h-pyrazinecarboxylate, 4-4-boc-piperazinyl phenylboronic acid pinacol ester, 4-n-tert-butoxycarbonylpiperazine benzeneboronic acid pinacol ester, tert-butyl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazine-1-carboxylate
CC(C)(C)OC(=O)N1CCN(CC1)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
388.32
388.31
90%
MFCD06411544
ZMAVVXGWEHZLDW-UHFFFAOYSA-N
tert-butyl 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazine-1-carboxylate
2795514
tert-butyl 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl) phenyl]piperazine-1-carboxylate
Videos
SDS
Documents

Documents

Product Certifications