missing translation for 'onlineSavingsMsg'
Learn More

T-box 19 Antibody (OTI1C9), Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18106006 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18106006 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18106006 Supplier Novus Biologicals Supplier No. NBP274469AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

T-box 19 Monoclonal specifically detects T-box 19 in Human, Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen T-box 19
Applications Western Blot
Classification Monoclonal
Clone OTI1C9
Conjugate Alexa Fluor 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias dJ747L4.1, FLJ26302, FLJ34085, T-box 19, T-box factor, pituitary, T-box protein 19, T-box transcription factor TBX19, TBS 19, TBS19, TPITFLJ34543
Gene Symbols TBX19
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-238 of human T-box 19 (NP_005140) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 9095.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.