missing translation for 'onlineSavingsMsg'
Learn More
Learn More
T-bet/TBX21 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41235-0.025mg
This item is not returnable.
View return policy
Description
T-bet/TBX21 Polyclonal specifically detects T-bet/TBX21 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| T-bet/TBX21 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| TBX21 | |
| Antibody was raised against a 19 amino acid peptide near the carboxy terminus of human TBX2. The immunogen is located within the last 50 amino acids of TBX21 . Amino Acid Squence: APSPFDKEAEGQFYNYFPN | |
| Affinity Purified | |
| RUO | |
| 30009 | |
| Human, Mouse | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence | |
| T-bet, TBETT-PET, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, Transcription factor TBLYM | |
| Rabbit | |
| 55 kDa | |
| 0.025 mg | |
| Primary | |
| TBX21 antibody is human and mouse reactive. | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction