missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ T-bet Recombinant Rabbit Monoclonal Antibody (9M7X6)
Rabbit Recombinant Monoclonal Antibody
£147.00 - £363.00
Specifications
| Antigen | T-bet |
|---|---|
| Clone | 9M7X6 |
| Concentration | 1.7 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Immunoprecipitation, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30284242
|
Invitrogen™
MA558223 |
20 μL |
£147.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30284559
|
Invitrogen™
MA558224 |
100 μL |
£363.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: KGFRENFESM YTSVDTSIPS PPGPNCQFLG GDHYSPLLPN QYPVPSRFYP DLPGQAKDVV PQAYWLGAPR DHSYEAEFRA VSMKPAFLPS APGPTMSYYR GQEVLAPGAG WPVAPQYPPK MGPASWFRPM RTLPMEPGPG GSEGRGPEDQ GPPLVWTEIA PIRPESSDSG LGEGDSKRRR VSPYPSSGDS SSPAGAPSPF DKEAEGQFYN YFPN.
T-bet, also known as TBX21, is a T-cell specific transcription factor containing a single T-box DNA-binding domain. It controls the expression of the TH1 cytokine, interferon-gamma. Both Th1 and Th17 cells are crucial in immune regulation and autoimmune disease development. T-bet initiates Th1 lineage development from naive Th precursor cells and directs T cell differentiation to Th1 versus Th17. T-bet is cooperating with Stat4 (signal tranducer and activator of transcription 4) in programming chromatin architecture for Th1 gene expression. Genetic variations in T-bet are associated with susceptibility to aspirin-induced asthma and nasal polyps.Specifications
| T-bet | |
| 1.7 mg/mL | |
| ELISA, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q9UL17 | |
| 30009 | |
| Recombinant protein (or fragment). | |
| Antibody |
| 9M7X6 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| Tbx21 | |
| HGNC:11599; LOW QUALITY PROTEIN: T-box transcription factor TBX21; Tbet; T-bet; TBLYM; T-box 21; T-box expressed in T cells; T-box protein 21; T-box transcription factor 21; T-box transcription factor TBX21; TBT1; TBX21; T-cell-specific T-box transcription factor T-bet; T-PET; transcription factor TBLYM; transcription factor T-box 21 | |
| Tbx21 | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title