Learn More
Invitrogen™ T-bet Monoclonal Antibody (eBio4B10 (4B10)), PE, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ 12-5825-80
Description
Description: The eBio4B10 monoclonal antibody reacts with mouse, rhesus moneky and human T-bet. T-bet is a Th1-specific T-box transcription factor critical to the development of the Th1 CD4+ T cell lineage. This is known based on the observations that T-Bet deficient mice have impaired Th1 cell development, and that ectopic expression of T-Bet results in development skewed to the Th1 lineage. T-Bet expression is induced by the Th1 cytokine IFN gamma, and T-Bet also regulates the expression of IFN gamma, likely, at least in part, through the modification of DNA accessibility and histone remodeling. In addition to IFN gamma, T-Bet is also known to regulate the expression of IL-12R beta and IL-2. Moreover, T-Bet plays a role in class-switch recombination in B-cells. Applications Reported: This eBio4B10 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This eBio4B10 antibody has been tested by intracellular staining and flow cytometric analysis of normal human peripheral blood cells using the Foxp3/Transcription Factor Staining Buffer Set (cat. 00-5523) and protocol. This antibody can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
T-bet, also known as TBX21, is a T-cell specific transcription factor containing a single T-box DNA-binding domain. It controls the expression of the TH1 cytokine, interferon-gamma. Both Th1 and Th17 cells are crucial in immune regulation and autoimmune disease development. T-bet initiates Th1 lineage development from naive Th precursor cells and directs T cell differentiation to Th1 versus Th17. T-bet is cooperating with Stat4 (signal tranducer and activator of transcription 4) in programming chromatin architecture for Th1 gene expression. Genetic variations in T-bet are associated with susceptibility to aspirin-induced asthma and nasal polyps.
Specifications
| T-bet | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Q9JKD8, Q9UL17 | |
| Tbx21 | |
| Affinity chromatography | |
| RUO | |
| 30009, 57765, 694044 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| eBio4B10 (4B10) | |
| PE | |
| Tbx21 | |
| HGNC:11599; LOW QUALITY PROTEIN: T-box transcription factor TBX21; Tbet; T-bet; TBLYM; T-box 21; T-box expressed in T cells; T-box protein 21; T-box transcription factor 21; T-box transcription factor TBX21; TBT1; TBX21; T-cell-specific T-box transcription factor T-bet; T-PET; transcription factor TBLYM; transcription factor T-box 21 | |
| Mouse | |
| 25 μg | |
| Primary | |
| Human, Mouse, Rhesus Macaque | |
| Antibody | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.