missing translation for 'onlineSavingsMsg'
Learn More

T 26c disodium salt

Product Code. 18735726
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18735726 25 mg 25mg
18788396 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18735726 Supplier Tocris Bioscience Supplier No. 6134/25

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Highly potent and selective MMP13 inhibitor

T 26c disodium salt is a highly potent and selective MMP13 inhibitor (IC50 = 6.9 pM). Exhibits >2600-fold selectivity for MMP13 over related MMPs. Inhibits degradation of bovine nasal septum cartilage explants in vitro . Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Disodium 4-[[[[2-[[[[3-(Methyloxy)phenyl]methyl]amino]carbonyl]-4-oxo-3,4-dihydrothieno[2,3-d]pyrimidin-5-yl]methyl]oxy]methyl]benzoate
CAS 869298-22-6
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C24H19N3Na2O6S
Formula Weight Observed MW: 523.47
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.