missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ T-2 Mycotoxin Monoclonal Antibody (4A6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549615
This item is not returnable.
View return policy
Description
Antibody tested in indirect ELISA.
T-2 mycotoxin is a toxic secondary metabolite produced by several species of fungi, including Fusarium sporotrichioides and Fusarium poae. These fungi can grow on a variety of crops, including wheat, barley, oats, and corn, and can produce T-2 mycotoxin under certain conditions. T-2 mycotoxin is a potent inhibitor of protein synthesis and can cause a variety of toxic effects in animals, including vomiting, diarrhea, hemorrhaging, and immunosuppression. It is also a potential human carcinogen and has been linked to esophageal cancer in some populations.
Specifications
| T-2 Mycotoxin | |
| Monoclonal | |
| 4.1 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Mouse | |
| Protein G | |
| RUO | |
| Fungi | |
| Antibody | |
| IgG2b |
| ELISA, Immunohistochemistry, Lateral Flow, SDS-Page | |
| 4A6 | |
| Unconjugated | |
| Mycotoxin T2; Mycotoxin T-2; T2 Toxin; T-2 Toxin | |
| Conjugate of T-2 hapten and KLH. | |
| 200 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction