Learn More
SYVN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13473-1-AP-150UL
 View more versions of this product
Product Code. 16871704
  • £349.00 / 150µL
Description
HRD1 is a ubiquitin ligase whose expression is induced by the unfolded protein response (UPR) following endoplasmic reticulum stress. Expression of HRD1 protects cells from apoptosis by inducing degradation of abnormally processed proteins that accumulate in the endoplasmic reticulum. HRD1 is expressed in many tissues, strongly expressed in brain, pancreas, liver, kidney and skeletal muscle. Amano T, et al. reported that Synoviolin/Hrd1 (expressed in rheumatoid synovium) is a novel causative factor for arthropathy by triggering synovial cell outgrowth through its antiapoptotic effects. HRD1 contains one ring-type zinc finger.
Specifications
Specifications
SYVN1
Polyclonal
Unconjugated
SYVN1
HRD1, KIAA1810, Synovial apoptosis inhibitor 1, SYVN1
Rabbit
Antigen Affinity Chromatography
RUO
361712, 74126, 84447
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q86TM6, Q9DBY1
Syvn1
SYVN1 Fusion Protein Ag4267
150 μL
Primary
Human, Mouse, Rat