missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SYTL5 Polyclonal Antibody

Description
A suggested positive control is Hela cell lysate. PA5-21176 can be used with blocking peptide PEP-1290.
SYTL5 belongs to the synaptotagmin-like (Slp) protein family, which contains a unique Slp homolog domain (SHD) at the N-terminus. The SHD functions as a binding site for Rab27A, which plays a role in protein transport. Expression of SYTL5 is restricted to placenta and liver, suggesting that it might be involved in Rab27A-dependent membrane trafficking in specific tissues. Several members of the synaptotagmin family are calcium-sensing vesicle-fusion proteins as mediators of cell migration. SYTL7 & SYTL5 were positive regulators of chemotaxis, whereas SYTL2 was a negative regulator of chemotaxis.
Specifications
Specifications
| Antigen | SYTL5 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | SYTL5 |
| Gene Accession No. | Q80T23, Q812E4, Q8TDW5 |
| Gene Alias | ENSMUSG00000054453; exophilin 9; Slp5; synaptotagmin 15; synaptotagmin like 5; synaptotagmin-like 5; synaptotagmin-like protein 5; Sytl5 |
| Gene Symbols | SYTL5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?