missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SYTL4 Monoclonal Antibody (OTI2B3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527324
This item is not returnable.
View return policy
Description
SYTL4 Monoclonal Antibody for Western Blot, IHC (P)
This gene encodes a member of the synaptotagmin like protein family. Members of this family are characterized by an N-terminal Rab27 binding domain and C-terminal tandem C2 domains. The encoded protein binds specific small Rab GTPases and is involved in intracellular membrane trafficking. This protein binds Rab27 and may be involved in inhibiting dense core vesicle exocytosis. Alternate splicing results in multiple transcript variants that encode the same protein.
Specifications
| SYTL4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96C24 | |
| SYTL4 | |
| Full length human recombinant protein of SYTL4 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2B3 | |
| Unconjugated | |
| SYTL4 | |
| Exophilin-2; Gph; Granuphilin; granuphilin b; granuphilin-a; granuphilin-b; Slp4; synaptotagmin like 4; synaptotagmin-like 4; synaptotagmin-like 4 (granuphilin-a); synaptotagmin-like protein 4; Sytl4 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 94121 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction