missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYTL3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22076-1-AP-20UL
This item is not returnable.
View return policy
Description
May act as Rab effector protein and play a role in vesicle trafficking. Binds phospholipids in the presence of calcium ions.Specifications
| SYTL3 | |
| Polyclonal | |
| Unconjugated | |
| SYTL3 | |
| Exophilin 6, SLP3, synaptotagmin like 3, Synaptotagmin like protein 3, SYTL3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 499017, 83672, 94120 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q4VX76, Q99N48 | |
| Sytl3 | |
| SYTL3 Fusion Protein Ag17219 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction