missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYTL1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA558126

Product Code. 15756011

  • £437.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: EAIAGVLQRD ARLRQLEEGR VSKLRASVAD PGQLKILTGD WFQEARSQRH HNAHFGSDLV RASMRRKKST RGDQAPGHDR EAEAAVKEK Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 79%.

SYTL1 may play a role in vesicle trafficking. Binds phosphatidylinositol 3,4,5-trisphosphate. Acts as a RAB27A effector protein and may play a role in cytotoxic granule exocytosis in lymphocytes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SYTL1
Polyclonal
Unconjugated
SYTL1
exophilin-7; FLJ14996; JFC1; NADPH oxidase-related, C2 domain-containing protein; Protein JFC1; Protein SLP1; SB146; SLP1; synaptotagmin like 1; synaptotagmin-like 1; synaptotagmin-like protein 1; SYTL1
Rabbit
Antigen Affinity Chromatography
RUO
84958
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.6 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8IYJ3
SYTL1
Recombinant protein corresponding to Human SYTL1. Recombinant protein control fragment (Product #RP-94260).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.