missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synaptotagmin 4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550269
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total SYT4 protein.
Synaptotagmin 4 Polyclonal antibody specifically detects Synaptotagmin 4 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
Synaptotagmin 4 | |
Polyclonal | |
Unconjugated | |
Syt4 | |
HsT1192; KIAA1342; synaptotagmin 4; synaptotagmin IV; Synaptotagmin4; Synaptotagmin-4; Syt3; Syt4; SytIV | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6860 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9H2B2 | |
Syt4 | |
Fusion protein corresponding to a region derived from internal residues of human synaptotagmin IV | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |