missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYT2 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £295.00
Specifications
| Antigen | SYT2 |
|---|---|
| Concentration | 550 μg/mL |
| Applications | Western Blot, Immunofluorescence, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30233256
|
Proteintech
21245-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30233423
|
Proteintech
21245-1-AP-150UL |
150 μL |
£349.00 £295.00 / 150µL Save £54.00 15% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SYT2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SYT2. This antibody recognizes human, mouse, rat antigen. The SYT2 antibody has been validated for the following applications: WB, IF-P, ELISA analysis.Specifications
| SYT2 | |
| Western Blot, Immunofluorescence, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| FLJ42519; SytII | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
| 550 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 127833 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title