Learn More
SYT12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA544988
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: AEAREKRVPA WNAQRASTRG PPSRKGSLSI EDTFESISEL GPLELMGREL Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 92%; Horse: 93%; Human: 100%; Mouse: 86%; Pig: 100%; Rabbit: 93%; Rat: 86%
SYT12 Polyclonal antibody specifically detects SYT12 in Human samples. It is validated for Western Blot.Specifications
SYT12 | |
Polyclonal | |
Unconjugated | |
SYT12 | |
Srg1; synaptotagmin 12; synaptotagmin XII; Synaptotagmin12; Synaptotagmin-12; synaptotagmin-related gene 1 protein; SynaptotagminXII; synaptotagmin-XII; Syt XII; SYT11; Syt12; SYT-12; Sytr1; SytXII | |
Rabbit | |
Affinity Chromatography | |
RUO | |
91683 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8IV01 | |
SYT12 | |
Synthetic peptide directed towards the N-terminal of human SYT12 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |