missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SYT11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106754
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SYT11.
This gene is a member of the synaptotagmin gene family and encodes a protein similar to other family members that are known calcium sensors and mediate calcium-dependent regulation of membrane trafficking in synaptic transmission. The encoded protein is also a substrate for ubiquitin-E3-ligase parkin. The gene has previously been referred to as synaptotagmin XII but has been renamed synaptotagmin XI to be consistent with mouse and rat official nomenclature. [provided by RefSeq, Apr 2010].
Specifications
| SYT11 | |
| Polyclonal | |
| Unconjugated | |
| SYT11 | |
| 1500004A13Rik; 3632445O20Rik; 5430404N14Rik; 6530420C11Rik; AI851970; KIAA0080; synaptotagmin 11; synaptotagmin 12; synaptotagmin XI; Synaptotagmin-11; SYT11; SYT12; SytXI | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 229521, 23208, 60568 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O08835, Q9BT88, Q9R0N3 | |
| SYT11 | |
| A synthesized peptide derived from human SYT11(Accession Q9BT88), corresponding to amino acid residues H182-I232. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction