missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SYT10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563126
This item is not returnable.
View return policy
Description
Immunogen sequence: MSFHKEDGVN SLCQKALHIV TELCFAGQVE WEKCSGIFPR DRGSQGGSST DIS Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.
SYT10 is a Ca2+ sensor specifically required for the Ca2+-dependent exocytosis of secretory vesicles containing IGF1 in neurons of the olfactory bulb. Exocytosis of IGF1 is required for sensory perception of smell. SYT10 is not involved in Ca2+-dependent synaptic vesicle exocytosis. It acts through Ca2+ and phospholipid binding to the C2 domain: Ca2+ induces binding of the C2-domains to phospholipid membranes and to assembled SNARE-complexes; both actions contribute to triggering exocytosis.
Specifications
| SYT10 | |
| Polyclonal | |
| Unconjugated | |
| SYT10 | |
| DKFZp781D2042; P65; SVP65; synaptotagmin 10; synaptotagmin X; Synaptotagmin-10; SYT; Syt10; SYT-10; SytX | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 341359 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6XYQ8 | |
| SYT10 | |
| Recombinant protein corresponding to Human SYT10. Recombinant protein control fragment (Product #RP-101200). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction