missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SYT10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570844
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 79%; Dog: 86%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 79%; Rabbit: 86%; Rat: 86%.
SYT10 is a Ca2+ sensor specifically required for the Ca2+-dependent exocytosis of secretory vesicles containing IGF1 in neurons of the olfactory bulb. Exocytosis of IGF1 is required for sensory perception of smell. SYT10 is not involved in Ca2+-dependent synaptic vesicle exocytosis. It acts through Ca2+ and phospholipid binding to the C2 domain: Ca2+ induces binding of the C2-domains to phospholipid membranes and to assembled SNARE-complexes; both actions contribute to triggering exocytosis.
Specifications
| SYT10 | |
| Polyclonal | |
| Unconjugated | |
| SYT10 | |
| DKFZp781D2042; P65; SVP65; synaptotagmin 10; synaptotagmin X; Synaptotagmin-10; SYT; Syt10; SYT-10; SytX | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 341359 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6XYQ8 | |
| SYT10 | |
| synthetic peptide directed towards the middle region of human SYT10. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction