Learn More
CELLPATH SYSTEM II+ MICRO BIOPSY CASSETTE
Click to view available options
Material:
Hoppered
Loose
Threaded
Unit Size:
Pack of 1000
Pack of 2000
Pack of 3000
Description
- Read rates
- Robust and secure closure ensures specimen security
- Designed to improve reagent flow
- Large color range
- Writing surface suitable for marking with a CellMark pen
Specifications
Specifications
| Color | White |
| Material | Threaded |
| Product Type | Cassette |