missing translation for 'onlineSavingsMsg'
Learn More

Syringic acid, MedChemExpress

Product Code. 16459056
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16459056 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16459056 Supplier Medchem Express Supplier No. HYN0339
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Syringic acid is correlated with high antioxidant activity and inhibition of LDL oxidation.

Syringic acid is correlated with high antioxidant activity and inhibition of LDL oxidation.

Specifications

Chemical Name or Material Syringic acid
CAS 530-57-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C9H10O5
Quantity 100 mg
Solubility Information DMSO : 62.5 mg/mL (315.39 mM; Need ultrasonic)
SMILES O=C(O)C1=CC(OC)=C(O)C(OC)=C1
Molecular Weight (g/mol) 198.17
Formula Weight 198.17
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.