missing translation for 'onlineSavingsMsg'
Learn More

Syringic acid, 97%

Product Code. 11498970
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498970 25 g 25g
11408980 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11498970 Supplier Thermo Scientific Alfa Aesar Supplier No. A11725.14
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 530-57-4 | C9H10O5 | 198.17 g/mol

Syringic acid is used as an inhibitor of poly(ethylene glycol) oxidation. It is used for the preparation of poly(1,4-phenylene oxide). It is added as an additive in wine and vinegar.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Syringic acid is used as an inhibitor of poly(ethylene glycol) oxidation. It is used for the preparation of poly(1,4-phenylene oxide). It is added as an additive in wine and vinegar.

Solubility
Soluble in methanol (50 mg/ml), water (5.78 mg/ml at 25°C), chloroform: methanol (1:1), ethyl alcohol, and DMF.

Notes
Storage temperature: 2 - 4°C. Avoid excessive heat and light. Air and moisture sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 530-57-4
Molecular Formula C9H10O5
Molecular Weight (g/mol) 198.17
MDL Number MFCD00002552
InChI Key JMSVCTWVEWCHDZ-UHFFFAOYSA-N
Synonym syringic acid, 3,5-dimethoxy-4-hydroxybenzoic acid, cedar acid, gallic acid 3,5-dimethyl ether, benzoic acid, 4-hydroxy-3,5-dimethoxy, 3,5-dimethoxy-4-hydroxybenzyl acid, 4-hydroxy-3,5-dimethoxy-benzoic acid, chembl1414, syringicacid, syringlicacid
PubChem CID 10742
ChEBI CHEBI:68329
IUPAC Name 4-hydroxy-3,5-dimethoxybenzoic acid
SMILES COC1=CC(=CC(OC)=C1O)C(O)=O

Specifications

Melting Point 206°C to 208°C
Boiling Point 192°C to 193°C (14 mmHg)
Quantity 25 g
Beilstein 2115262
Solubility Information Soluble in methanol (50mg/ml),water (5.78mg/ml at 25°C),chloroform: methanol (1:1),ethyl alcohol,and DMF.
Formula Weight 198.17
Percent Purity ≥98%
Chemical Name or Material Syringic acid
compliance-icons
Product Identifier
  • Syringic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Syringic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.