missing translation for 'onlineSavingsMsg'
Learn More

Syringic acid, 97%

Product Code. 10654612 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10654612 10 g Glass bottle 10g
10765791 25 g Glass bottle 25g
10628634 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10654612 Supplier Thermo Scientific Acros Supplier No. 132890100
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 530-57-4 | C9H10O5 | 198.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 530-57-4
Molecular Formula C9H10O5
Molecular Weight (g/mol) 198.17
MDL Number MFCD00002552
InChI Key JMSVCTWVEWCHDZ-UHFFFAOYSA-N
Synonym syringic acid, 3,5-dimethoxy-4-hydroxybenzoic acid, cedar acid, gallic acid 3,5-dimethyl ether, benzoic acid, 4-hydroxy-3,5-dimethoxy, 3,5-dimethoxy-4-hydroxybenzyl acid, 4-hydroxy-3,5-dimethoxy-benzoic acid, chembl1414, syringicacid, syringlicacid
PubChem CID 10742
ChEBI CHEBI:68329
IUPAC Name 4-hydroxy-3,5-dimethoxybenzoic acid
SMILES COC1=CC(=CC(OC)=C1O)C(O)=O

Specifications

Melting Point 206°C to 212°C
Color Beige-Gray to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 10 g
Beilstein 10, 480
Solubility Information Solubility in water: 5780mg/l (25°C)
Formula Weight 198.17
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Syringic acid
Show More Show Less
compliance-icons
Product Identifier
  • Syringic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.