Learn More
Hamilton™ Custom Cemented Needle Syringes
The gas-tight version of the original 700 series syringes.
Brand: Hamilton™ 82525
Code : NEW
Additional Details : Weight : 0.14500kg
Description
- Ideal for dispensing volumes from 1 up to 500 μL
- A variety of volumes, lengths, and gauges available
Specifications
Not Autoclavable | |
1025 | |
25 mL |
Syringe Needle | |
1 Ea. |