missing translation for 'onlineSavingsMsg'
Learn More

Pinnacle Brush™ Syringe Brushes

Features nylon bristles. Pinnacle Brush™ Syringe Brushes are for use with syringes

Brand:  Pinnacle Brush™ F1011/3

Code : 03

Additional Details : Weight : 0.03060kg

 View more versions of this product

Product Code. 12831954

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Nylon bristles
  • Fan shaped end
Specifications

Specifications

Syringe Brushes
5mL Syringes
155 mm
Nylon
5mL Syringes
5 Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers