missing translation for 'onlineSavingsMsg'
Learn More

Pinnacle Brush™ Syringe Brushes

Features nylon bristles. Pinnacle Brush™ Syringe Brushes are for use with syringes

Brand:  Pinnacle Brush™ F1011/5

Code : 03

Additional Details : Weight : 0.08640kg

 View more versions of this product

Product Code. 12398328

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Nylon bristles
  • Fan shaped end
Specifications

Specifications

Syringe Brushes
100mL Syringes
255 mm
Nylon
100mL Syringes
5 Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers