missing translation for 'onlineSavingsMsg'
Learn More

Syringaldehyde, MedChemExpress

Product Code. 16459156
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16459156 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16459156 Supplier Medchem Express Supplier No. HYN1390

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Syringaldehyde is a polyphenolic compound belonging to the group of flavonoids and is found in different plant species like Manihot esculenta and Magnolia officinalis. Syringaldehyde moderately inhibits COX-2 activity with an IC50 of 3.5 μg/mL. Anti-hyperglycemic and anti-inflammatory activities.

Syringaldehyde is a polyphenolic compound belonging to the group of flavonoids and is found in different plant species like Manihot esculenta and Magnolia officinalis. Syringaldehyde moderately inhibits COX-2 activity with an IC50 of 3.5 μg/mL. Anti-hyperglycemic and anti-inflammatory activities.

Specifications

Chemical Name or Material Syringaldehyde
CAS 134-96-3
Color Off-White to Brown
Purity Grade Notes Research
Molecular Formula C9H10O4
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (548.94 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=CC1=CC(OC)=C(O)C(OC)=C1
Molecular Weight (g/mol) 182.17
Formula Weight 182.17
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.