missing translation for 'onlineSavingsMsg'
Learn More

Syringaldehyde, 98+%

Product Code. 11499020
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499020 5 g 5g
11409030 25 g 25g
11419030 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499020 Supplier Thermo Scientific Alfa Aesar Supplier No. A12933.06
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 134-96-3 | C9H10O4 | 182.18 g/mol

Syringaldehyde has bioactive properties and is, therefore, used in pharmaceuticals, food, cosmetics, textiles, pulp and paper industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Syringaldehyde has bioactive properties and is, therefore, used in pharmaceuticals, food, cosmetics, textiles, pulp and paper industries.

Solubility
Very sparingly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Store at room temperature. It is sensitive to air. Incompatible with strong oxidizing agents, and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134-96-3
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00006943
InChI Key KCDXJAYRVLXPFO-UHFFFAOYSA-N
Synonym syringaldehyde, 3,5-dimethoxy-4-hydroxybenzaldehyde, syringic aldehyde, syringylaldehyde, syringealdehyde, benzaldehyde, 4-hydroxy-3,5-dimethoxy, gallaldehyde 3,5-dimethyl ether, 3,5-dimethoxy-4-hydroxybenzene carbonal, 4-hydroxy-3,5-dimethoxy-benzaldehyde, springaldehyde
PubChem CID 8655
ChEBI CHEBI:67380
IUPAC Name 4-hydroxy-3,5-dimethoxybenzaldehyde
SMILES COC1=CC(C=O)=CC(OC)=C1O

Specifications

Melting Point 110°C to 113°C
Boiling Point 192°C to 193°C (14 mmHg)
Quantity 5 g
Beilstein 784514
Sensitivity Air Sensitive
Merck Index 14,9015
Solubility Information Very sparingly soluble in water.
Formula Weight 182.18
Percent Purity ≥98%
Chemical Name or Material Syringaldehyde
compliance-icons
Product Identifier
  • Syringaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Syringaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.