missing translation for 'onlineSavingsMsg'
Learn More

Syringaldazine 99+%

Product Code. 16564194 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16564194

Brand: Thermo Scientific Acros 175930010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14414-32-5 | C18H20N2O6 | 360.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14414-32-5
Molecular Formula C18H20N2O6
InChI Key MYNVJNZGTAVRJD-UHFFFAOYSA-N
Synonym syringaldazine, 4-hydroxy-3,5-dimethoxybenzaldehyde azine, 3,5-dimethoxy-4-hydroxybenzalazine, 4,4'-hydrazine-1,2-diylidenebis methanylylidene bis 2,6-dimethoxyphenol, benzaldehyde, 4-hydroxy-3,5-dimethoxy-, 4-hydroxy-3,5-dimethoxyphenyl methylene hydrazone, 4-1e-e-2-4-hydroxy-3,5-dimethoxyphenyl methylidene hydrazin-1-ylidene methyl-2,6-dimethoxyphenol, 4-2-4-hydroxy-3,5-dimethoxyphenyl methylidene hydrazin-1-ylidene methyl-2,6-dimethoxyphenol, benzaldehyde, 4-hydroxy-3,5-dimethoxy-, 2-4-hydroxy-3,5-dimethoxyphenyl methylene hydrazone
PubChem CID 5379425

Specifications

Melting Point 209°C to 213°C
Color Green to Yellow
Infrared Spectrum Authentic
Quantity 1 g
SMILES COC1=CC(=CNNC=C2C=C(C(=O)C(=C2)OC)OC)C=C(C1=O)OC
IUPAC Name 4-[[2-[(3,5-dimethoxy-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]hydrazinyl]methylidene]-2,6-dimethoxycyclohexa-2,5-dien-1-one
Molecular Weight (g/mol) 360.366
Formula Weight 360.36
Percent Purity ≥99.0%
Physical Form Fine Crystalline Powder
Chemical Name or Material Syringaldazine, 99+%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.